Disodium anthraquinone-2,6-disulfonate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 853-68-9
MDL Number: MFCD00001230
Molecular Formula: C14H8O8S2
Molecular Weight: 412.29
Purity/Analysis Method:
Other Product Names / Synonyms: Anthraquinone-2,6-disulfonic Acid Disodium Salt
Formula:
C₁₄H₈O₈S₂ |
MDL Number:
MFCD00001230 CAS Number: 853-68-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...