Acetonitrile ≥99.5% (by GC) for spectrophotometry
Supplier: TCI America
|
Danger
|
Synonyms:
Methanecarbonitrile, Methane, cyano-, Methyl cyanide, Ethyl nitrile, ACN, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
A0293-100ML
TCA0293-100MLEA
56.2
USD
TCA0293-100ML
Acetonitrile ≥99.5% (by GC) for spectrophotometry
Acetonitrile
[for Spectrophotometry]
CAS Number: 75-05-8
MDL Number: MFCD00001878
Molecular Formula: C2H3N
Molecular Weight: 41.05
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Melting point (°C): -46
Flash Point (°C): 10
Specific Gravity (20/20): 0.78
Product Categories: Analytical Science, Solvents for Spectrophotometry
Other Product Names / Synonyms: Methyl Cyanide, ACN
Formula:
H₃CCN MW: 41.05 g/mol Boiling Pt: 82 °C Melting Pt: –46 °C Density: 0.78 (20 °C) Flash Pt: 10 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II REACH: 01-2119471307-38 Merck Index: 13,00071 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...