2-Amino-4-chloro-5-methylbenzenesulfonic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-Amino-2-chlorotoluene-5-sulphonic acid, 4-Amino-2-chlorotoluene-5-sulfonic acid

A0286-25G
TCA0286-025GEA 29 USD
TCA0286-025G
2-Amino-4-chloro-5-methylbenzenesulfonic acid ≥98.0% (by HPLC, titration analysis)
2-Amino-4-chloro-5-methylbenzenesulphonic acid

CAS Number: 88-51-7 MDL Number: MFCD00035771 Molecular Formula: C7H8ClNO3S Molecular Weight: 221.66 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal


Other Product Names / Synonyms: 5-Chloro-4-methylaniline-2-sulfonic Acid, 5-Chloro-p-toluidine-2-sulfonic Acid, 2-Amino-4-chloro-5-methylbenzenesulfonic Acid

Formula: C₇H₈ClNO₃S
MW: 221.6613 g/mol
MDL Number: MFCD00035771
CAS Number: 88-51-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50f9991ee333269aaf0f4f25b870affbdbf2ec8a&vid=50f9991ee333269aaf0f4f25b870affbdbf2ec8a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR