Isoamyl acetate ≥98.0%

Supplier: TCI America
Danger

Synonyms: Isopentyl acetate, Acetic acid isoamyl ester

A0264-100ML
TCA0264-100MLEA 76 USD
TCA0264-100ML
Isoamyl acetate ≥98.0%
Isoamyl acetate

[for Spectrophotometry] CAS Number: 123-92-2 MDL Number: MFCD00008946 Molecular Formula: C7H14O2 Molecular Weight: 130.19 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 142 Flash Point (°C): 38 Specific Gravity (20/20): 0.87


Product Categories: Bioscience, Treatment Agents (Staining), Analytical Science, Solvents for Spectrophotometry Other Product Names / Synonyms: Acetic Acid Isoamyl Ester, 3-Methylbutyl Acetate, Isopentyl Acetate

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 142 °C
Melting Pt: –78 °C
Density: 0.87 (20 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00008946
CAS Number: 123-92-2
EINECS: 204-662-3
UN: 1104
ADR: 3,III
Merck Index: 12,05125
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=764f4f5d7d57e064b44b38d125ff76e4c49e6ab7&vid=221bda2b81602615c31dce7ef06cb38db2e4a057&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR