Acrylamide polymer, M.W. M.W.=400,000-800,000 10% in water (containing small amounts of formalin as fungicide)
Supplier: TCI America
A0140-500G
TCA0140-500GEA
208.34
USD
TCA0140-500G
Acrylamide polymer, M.W. M.W.=400,000-800,000 10% in water (containing small amounts of formalin as fungicide)
Acrylamide polymer
CAS Number: 9003-05-8
MDL Number: MFCD00084392
Form: Starch Syrup
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Polymers
Formula:
(CH2:CHCONH2)n |
MDL Number:
MFCD00084392 CAS Number: 9003-05-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...