N-Acetylglycine(Ac-Gly-OH) ≥99.0%
Supplier: TCI America
Synonyms:
Ac-Gly-OH, N-Acetylglycine, Acetamidoacetic acid, Aceturic acid
A0093-25G
A0093-500G
A0093-100G
TCA0093-025GEA
21
USD
TCA0093-025G
TCA0093-500G
TCA0093-100G
N-Acetylglycine(Ac-Gly-OH) ≥99.0%
N-Acetylglycine (Ac-Gly-OH)
CAS Number: 543-24-8
MDL Number: MFCD00004275
Molecular Formula: C4H7NO3
Molecular Weight: 117.10
Purity/Analysis Method:
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Acetamidoacetic Acid, Aceturic Acid, Ac-Gly-OH
Formula:
C₄H₇NO₃ Melting Pt: 207 °C |
MDL Number:
MFCD00004275 CAS Number: 543-24-8 EINECS: 208-839-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...