4'-Acetoxyacetanilide ≥99.0% (by HPLC, total nitrogen)
Supplier: TCI America
CAS Number: 2623-33-8
MDL Number: MFCD00059205
Molecular Formula: C10H11NO3
Molecular Weight: 193.20
Purity/Analysis Method:
Other Product Names / Synonyms: N,O-Diacetyl-4-aminophenol
Formula:
C₁₀H₁₁NO₃ Melting Pt: 155 °C |
MDL Number:
MFCD00059205 CAS Number: 2623-33-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...