Acetonedimethyl acetal ≥98.0%

Supplier: TCI America
Danger

Synonyms: 2,2-Dimethoxypropane, Dimethoxypropane, DMP, Dimethoxy-2,2-propane

A0057-500ML A0057-25ML
TCA0057-500MLEA 49.28 USD
TCA0057-500ML TCA0057-25ML
Acetonedimethyl acetal ≥98.0%
Acetone-dimethyl acetal

CAS Number: 77-76-9 MDL Number: MFCD00008479 Molecular Formula: C5H12O2 Molecular Weight: 104.15 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 80 Flash Point (°C): -9 Specific Gravity (20/20): 0.85


Product Categories: Bioscience, Reagents for Oligosaccharide Synthesis Other Product Names / Synonyms: Acetone Dimethyl Acetal

Formula: C₅H₁₂O₂
Boiling Pt: 80 °C
Melting Pt: –47 °C
Density: 0.85 (20 °C)
Flash Pt: –9 °C
Storage Temperature: Ambient
MDL Number: MFCD00008479
CAS Number: 77-76-9
EINECS: 201-056-0
UN: 3271
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6936f6b50ad13ab1937b8330817793655823a65&vid=9b0a772e59fcf58b8b5469cf364cbb732b4c4bc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR