Dimethyl-3-oxoglutarate ≥95.0%

Supplier: TCI America

Synonyms: Dimethyl 1,3-acetonedicarboxylate, Dimethyl acetone-1,3-dicarboxylate

A0056-25G A0056-500G A0056-100G
TCA0056-25GEA 77 USD
TCA0056-25G TCA0056-500G TCA0056-100G
Dimethyl-3-oxoglutarate ≥95.0%
Dimethyl 3-oxoglutarate

CAS Number: 1830-54-2 MDL Number: MFCD00008462 Molecular Formula: C7H10O5 Molecular Weight: 174.15 Purity/Analysis Method: 95.0% (GC) Form: Clear Liquid Boiling point (°C): 150 Flash Point (°C): 110 Specific Gravity (20/20): 1.21


Other Product Names / Synonyms: 1,3-Acetonedicarboxylic Acid Dimethyl Ester, Dimethyl 3-Oxoglutarate, 3-Oxoglutaric Acid Dimethyl Ester, Dimethyl 3-Oxopentanedioate, 3-Oxopentanedioic Acid Dimethyl Ester

Formula: C₇H₁₀O₅
Boiling Pt: 150 °C
Density: 1.21 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008462
CAS Number: 1830-54-2
EINECS: 217-385-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f394fe04673efa6bdecab70493090ebcda6127a&vid=4f394fe04673efa6bdecab70493090ebcda6127a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR