Acetone ≥99.5%

Supplier: TCI America
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

A0054-500ML
TCA0054-500MLEA 65.07 USD
TCA0054-500ML
Acetone ≥99.5%
Acetone

[for Spectrophotometry]
CAS Number: 67-64-1
MDL Number: MFCD00008765
Molecular Formula: C3H6O
Molecular Weight: 58.08
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 56
Melting point (°C): -95
Flash Point (°C): -20
Specific Gravity (20/20): 0.79

Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56 °C
Melting Pt: –95 °C
Density: 0.79 (20 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR