Amyl acetate ≥99.0%

Supplier: TCI America
Danger

Synonyms: n-Pentyl acetate, Acetic acid amyl ester, Acetic acid pentyl ester, Pentyl acetate

A0021-500ML A0021-25ML
TCA0021-500MLEA 94.72 USD
TCA0021-500ML TCA0021-25ML
Amyl acetate ≥99.0%
Amyl acetate

CAS Number: 628-63-7 MDL Number: MFCD00009500 Molecular Formula: C7H14O2 Molecular Weight: 130.19 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 148 Flash Point (°C): 45 Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: Acetic Acid Amyl Ester, Pentyl Acetate

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 149
Melting Pt: -71
Density: 0.88 (20 °C)
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00009500
CAS Number: 628-63-7
EINECS: 211-047-3
UN: 1104
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcebb8adcd536407ec2a19a334d1dcbad6b0acec&vid=b96d23095d205e1a275f89c8c817ce1ee945d175&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR