Set of 3 Solutions for Bromide in H₂O, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
AS-BR9-SET
SXAS-BR9-SETEA 272.58 USD
SXAS-BR9-SET
Set of 3 Solutions for Bromide in H₂O, SPEX CertiPrep
Standards Anionic Standards Bromide Standards
Ion chromatography and ion selective electrode standards.
Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary. SPEX CertiPrep offers the highest quality IC standards available for the analytical laboratory.