Herculase II Fusion DNA Polymerases, Agilent Technologies
Supplier: AGILENT TECH
Herculase II Fusion DNA Polymerases produce superior yields with extension times as short as 15 sec/kb, even with genomic DNA targets up to 23kb. Herculase II helps overcome PCR challenges with successful amplification of targets of all complexity. Easily amplify low abundance, high GC, and other difficult targets with robust yields.