Dpn I, Restriction Enzymes, Agilent Technologies
Supplier: AGILENT TECH
500402
ST500402EA
145.52
USD
ST500402
Dpn I, Restriction Enzymes, Agilent Technologies
Nucleic Acid Reagents
Restriction Enzymes
Dpn I is an ideal enzyme for cloning and southern blot analysis.
- High purity enzyme preparation
Delivery information: Includes 10X universal reaction buffer.
Caution: For research use only. Not for use in diagnostic procedures.