Proteinase K
Supplier: AGILENT TECH
300140
ST300140EA
185.11
USD
ST300140
Proteinase K
Enzymes
Proteinase K is a broad spectrum protease from Tritirachium album that is ideal for preparing chromosomal DNA.
- For general digestion of protein in biological samples
- Lyophilized powder with specific activity >30 units/mg at 37 °C
It cleaves peptide bonds in which carbonyl groups are surrounded by aromatic, hydrophobic, or aliphatic amino acids.
Used for the following applications: pulsed field electrophoresis, chromosomal DNA for protein fingerprinting, and nuclease removal from DNA and RNA preparations.
Caution: For research use only. Not for use in diagnostic procedures.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)