Ampicillin, tablets, AmpTabs

Supplier: AGILENT TECH

300021
ST300021EA 448.33 USD
ST300021
Ampicillin, tablets, AmpTabs
Ampicillin

Amp Tabs Ampicillin Tablets are used to select for ampicillin-resistant bacteria and turbo Amp Antibiotic is used to reduce satellite colonies when selecting for ampicillin-resistant bacteria.


Caution: For Research Use Only. Not for Use in Diagnostic Procedures.

Formula: C₁₆H₁₉N₃O₄S
MW: 349.4 g/mol
MDL Number: MFCD00005175
CAS Number: 69-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR