Affinity Protein Expression and Purification Systems, Agilent Technologies
Supplier: AGILENT TECH
214303
99900-884EA 541.19 USD
99900-884 99900-886 99900-890 99900-892 ST214303
Affinity Protein Expression and Purification Systems, Agilent Technologies
Protein Expression Protein Expression Systems
The affinity protein expression and purification system provides a simple, gentle and effective method for high-level protein expression and purification.
  • pCAL vectors derived from T7 RNA polymerase-based pET vectors
  • Novel purification tag combines small size, gentle elution and high-affinity binding
  • Easy tag removal resulting in native protein
Delivery information: The complete system includes pCAL expression vectors containing a calmodulin-binding peptide tag and calmodulin affinity resin (available separately).
Caution: For research use only. Not for use in diagnostic procedures.