DYKDDDDK (FLAG®) Positive Control Lysate
Supplier: Rockland Immunochemical
W00-001-383
RLW00001383EA 194.33 USD
RLW00001383
DYKDDDDK (FLAG®) Positive Control Lysate
Antibodies
DYKDDDDK (FLAG®) Positive Control Lysate
  • Ready-to-use lysates
  • positive controls
This product is intended as a positive control for use with Rockland Immunochemicals anti-DYKDDDDK (FLAG®) tag antibodies.
Ready-to-use lysates are especially prepared as positive controls for separation by SDS-PAGE and subsequent western blot analysis. Lysates are prepared in denaturing buffer WITHOUT dissociating agents (i.e. no 2-mercaptoethanol or dithiothreitol has been added). If dissociating conditions are desired, add reducing agent prior to heating. Gently invert vial several times to ensure that the solution is homogeneous and that any precipitated material is re-dissolved. A loading volume of 0.25 to 2 µL is recommended for SDS-PAGE. The product is supplied in 1X loading buffer and is ready-to-use.