TMB Prestained Red ELISA Peroxidase Substrate, Rockland Immunochemicals
Supplier: Rockland Immunochemical
TMB Prestained Red ELISA Peroxidase Substrate
- pH: 4.0 +/- 0.2
- Stability @ 18° to 26°C: PASS
- Stability @ 4°C: PASS
- QC Raw Material: PASS
- Absorbance check of final product: PASS
- Performance Data per ELISA: PASS
TMB ELISA Peroxidase Substrate (3, 3', 5, 5' - Tetramethylbenzidine) is a chromogenic substrate used to visualize antibody reactivity in ELISA experiments. In the presence of peroxidases, TMB can act as an electron donor for the conversion of peroxides to water, typically changing the color of solution to a blue color equivalent to the degree of reactivity and upon addition of acid, changing the solution color to yellow. This proprietary formulation of TMB ELISA Peroxidase Substrate is red in color, enabling users to have visual certainty that the product was applied equally.
TMB ELISA Peroxidase Substrate is ideal for investigators in Immunology, Cancer, and Microbiology research.
TMB Prestained Red ELISA Peroxidase Substrate produces a soluble blue end product of ELISA, read at 370nm or 655nm.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...