Streptavidin, ATTO 647N
Supplier: Rockland Immunochemical
S000-56
RLS00056EA 266.42 USD
RLS00056
Streptavidin, ATTO 647N
Proteins and Peptides
STREPTAVIDIN ATTO 647N Conjugated
  • chromatographically pure Streptavidin
  • conjugated to ATTO 647N
  • No reaction was observed against anti-avidin
STREPTAVIDIN ATTO 647N is designed for STED microscopy, FRET, immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.
The emission spectra for this ATTO conjugate matches the principle output wavelengths of most common fluorescence instrumentation.