Streptavidin, ATTO 425
Supplier: Rockland Immunochemical
S000-51
RLS00051EA 266.42 USD
RLS00051
Streptavidin, ATTO 425
Proteins and Peptides
STREPTAVIDIN ATTO 425 Conjugated
  • chromatographically pure Streptavidin
  • conjugated to ATTO 425
  • No reaction was observed against anti-avidin
STREPTAVIDIN ATTO 425 is designed for STED microscopy, FRET, immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.
The emission spectra for this ATTO conjugate matches the principle output wavelengths of most common fluorescence instrumentation.