Rabbit Tissue, Rockland Immunochemicals
Supplier: Rockland Immunochemical
RB-T118 RB-T124 RB-T125 RB-T126 RB-T135 RB-T139 RB-T141 RB-T115 RB-T121 RB-T131 RB-T111 RB-T113 RB-T114 RB-T120 RB-T122 RB-T123 RB-T127 RB-T130 RB-T132 RB-T134 RB-T138 RB-T140 RB-T142 RB-T143 RB-T144 RB-T145 RB-T146 RB-T147 RB-T112 RB-T133
RLRB-T118EA 228.81 USD
RLRB-T118 RLRB-T124 RLRB-T125 RLRB-T126 RLRB-T135 RLRB-T139 RLRB-T141 RLRB-T115 RLRB-T121 RLRB-T131 RLRB-T111 RLRB-T113 RLRB-T114 RLRB-T120 RLRB-T122 RLRB-T123 RLRB-T127 RLRB-T130 RLRB-T132 RLRB-T134 RLRB-T138 RLRB-T140 RLRB-T142 RLRB-T143 RLRB-T144 RLRB-T145 RLRB-T146 RLRB-T147 RLRB-T112 RLRB-T133
Rabbit Tissue, Rockland Immunochemicals
Specimens Preserved Specimens Preserved Animal Materials
Use for DNA and RNA isolation.
  • Collected from antibiotic-free animals
  • DNA/RNA isolation
All tissues are collected from normal, healthy, mature animals that have been maintained on an antibiotic-free diet. Tissues are stored at −20°C or colder prior to use.
Ordering information: For specific breed, sex, or weight requirements, please contact your Avantor representative.