HUMAN BLOOD in Alsever's -, Rockland Immunochemicals
Supplier: Rockland Immunochemical
R314-0050
RLR314-0050EA 625 USD
RLR314-0050
HUMAN BLOOD in Alsever's -, Rockland Immunochemicals
Blood
HUMAN BLOOD in Alsevers -
  • aseptically collected and prepared to order
  • blood draw in Anticoagulant
Human whole blood is aseptically collected and processed upon receipt of your order. These cell suspensions are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Human red blood cells are perishable and are collected and processed upon receipt of your order.
Washed pooled cells preparations, complement titration, adsorption procedures, HA assays and for the preparation of stroma as particulate reagents.