Protein G,Biotin
Supplier: Rockland Immunochemical
PG00-06
RLPG00-06EA 292.54 USD
RLPG00-06
Protein G,Biotin
Proteins and Peptides
Protein G Biotin Conjugated
  • chromatographically pure Protein G
  • conjugated to Biotin
  • No reaction was observed against anti-Protein A
Protein G is a surface protein of two groups of Streptococcal bacteria that has the ability to bind immunoglobulins. Similar to Protein A, but with slightly different specificity, Protein G is an important agent in the purification of proteins due to its ability to bind the Fc region. Protein G also binds to albumin.
Protein G Biotin Conjugated is ideal for investigators involved in Signal Transduction, Molecular Biology, Cancer, and Cell Signaling.
Protein G Biotin Conjugated is suitable as a detection agent for primary antibodies that are of the IgG