Anti-Peroxidase (PAP) Soluble Complex Rat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemical
P400-0005
RLP400-0005EA 194.33 USD
RLP400-0005
Anti-Peroxidase (PAP) Soluble Complex Rat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Primary Rat Anti-Peroxidase (PAP) Soluble Complex Reacts with N/A

Type: Primary
Antigen: Peroxidase (PAP) Soluble Complex
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rat
Isotype:
Reactivity: None
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR