HDAC Assay Buffer, Rockland Immunochemicals
Supplier: Rockland Immunochemical
MB-068-0020
RLMB-068-0020EA 145.22 USD
RLMB-068-0020
HDAC Assay Buffer, Rockland Immunochemicals
Buffers Assay Buffers
Histone deacetylases (HDACs) are typically found in multiprotein complexes and are tightly regulated by subcellular localization, phosphorylation, and likely by other mechanisms.
  • 10X concentrated stock solution
  • (1X) 25 mM Tris Cl, 137 mM NaCl, 2.7 mM KCl and 1.0 mM MgCl2 at pH 8.0
  • Histone deacetylase Assays
HDAC assays allow the determination of deacetylase activity and provide accurate activity information that is reflective of endogenous regulation. HDAC Assay Buffer is typically employed with appropriate standards and controls.
Application Note: This product is a 10X concentrated stock solution.  Prepare a 1X working solution by diluting 1 part 10X concentrate with 9 parts distilled-deionized water or equivalent. Diluted (1X) HDAC Assay buffer consists of 25 mM Tris Cl, 137 mM NaCl, 2.7 mM KCl and 1.0 mM MgCl2 at pH 8.0.