Competent Cell Buffer, Rockland Immunochemicals
Supplier: Rockland Immunochemical
MB-057
RLMB-057EA 155.67 USD
RLMB-057
Competent Cell Buffer, Rockland Immunochemicals
Buffers Cloning Buffers
This product is ready-to-use as a working 1X solution and requires no further dilution
Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionised water.
This buffer consists of 60 mM Calcium Chloride, 15% v/v Glycerol, 10mM Tris HCl at a pH of 7.5
This product was aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.