Calcium chloride 0.5 mol/L (1 N) in aqueous solution

Supplier: Rockland Immunochemical

MB-055
RLMB-055EA 120.15 USD
RLMB-055
Calcium chloride 0.5 mol/L (1 N) in aqueous solution
Calcium chloride
Formula: CaCl₂
MDL Number: MFCD00010903
CAS Number: 10043-52-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR