Magnesium chloride 1 mol/L (2 N) in aqueous solution

Supplier: Rockland Immunochemical

MB-048
RLMB-048EA 130.6 USD
RLMB-048
Magnesium chloride 1 mol/L (2 N) in aqueous solution
Magnesium chloride
Formula: MgCl₂
MW: 95.21 g/mol
MDL Number: MFCD00011106
CAS Number: 7786-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR