Sodium chloride 5 mol/L (5 N) in aqueous solution

Supplier: Rockland Immunochemical

MB-047
RLMB-047EA 120.15 USD
RLMB-047
Sodium chloride 5 mol/L (5 N) in aqueous solution
Sodium chloride
Formula: NaCl
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR