SSPE Buffer, 20X concentrate, pH 7.4, Rockland Immunochemicals
Supplier: Rockland Immunochemical
MB-044
RLMB-044EA 161.94 USD
RLMB-044
SSPE Buffer, 20X concentrate, pH 7.4, Rockland Immunochemicals
Buffers Electrophoresis Buffers Hybridization Buffers
20X SSPE buffer (3.0 M Sodium Chloride, 0.2 M Sodium Hydrogen Phosphate, 0.02 M EDTA, pH 7.4) - DEPC treated.
This product was aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.