Epitope Tag Antibody Sampler Kit
Supplier: Rockland Immunochemical
Epitope tag antibody sampler kit detects overexpressed fusion proteins containing the corresponding epitope tags. DYKDDDDK Tag Antibody recognizes the DYKDDDDK peptide (the same epitope recognized by Sigma's Anti-FLAG® antibodies). Epitope tags are useful for the labeling and detection of proteins using western blotting, immunoprecipitation, and immunostaining techniques.
Contains: 6X HIS antibody, GFP Antibody, DYKDDDDK Antibody (Binds to same epitope as Sigma's Anti-FLAG® M2 Antibody), GST Antibody and Conjugated Secondary Antibodies
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...