Anti-Cyclin Dependent Kinase Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
K912
RLK912EA 1237.01 USD
RLK912
Anti-Cyclin Dependent Kinase Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-Cyclin Dependent Kinase Reacts with N/A

Type: Primary
Antigen: Cyclin Dependent Kinase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR