Guinea Pig Tissue, Rockland Immunochemicals
Supplier: Rockland Immunochemical
GP-T037 GP-T038 GP-T050 GP-T061 GP-T044 GP-T052 GP-T066 GP-T067 GP-T040 GP-T041 GP-T046 GP-T047 GP-T049 GP-T051 GP-T053 GP-T056 GP-T057 GP-T058 GP-T059 GP-T064 GP-T065 GP-T069 GP-T070 GP-T072 GP-T073 GP-T039 GP-T048 GP-T068 GP-T060 GP-T071
RLGP-T037EA 92.99 USD
RLGP-T037 RLGP-T038 RLGP-T050 RLGP-T061 RLGP-T044 RLGP-T052 RLGP-T066 RLGP-T067 RLGP-T040 RLGP-T041 RLGP-T046 RLGP-T047 RLGP-T049 RLGP-T051 RLGP-T053 RLGP-T056 RLGP-T057 RLGP-T058 RLGP-T059 RLGP-T064 RLGP-T065 RLGP-T069 RLGP-T070 RLGP-T072 RLGP-T073 RLGP-T039 RLGP-T048 RLGP-T068 RLGP-T060 RLGP-T071
Guinea Pig Tissue, Rockland Immunochemicals
Specimens Preserved Specimens Preserved Animal Materials
Use for DNA and RNA isolation.
  • Collected from antibiotic-free animals
  • DNA/RNA isolation
All tissues are collected from normal, healthy, mature animals that have been maintained on an antibiotic-free diet. Tissues are stored at −20°C or colder prior to use.
Ordering information: For specific breed, sex, or weight requirements, please contact your Avantor representative.