Sterile Monkey Plasma, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D516-04-2 D516-06-2
RLD516-04-2EA 939.25 USD
RLD516-04-2 RLD516-06-2
Sterile Monkey Plasma, Rockland Immunochemicals
Plasma
Plasma is obtained from non-hemolyzed blood collected from healthy, fasted donors
Animals are maintained on an antibiotic-free diet. Sterilization is accomplished by Millipore filtration.
Ordering information: For heat-inactivated sera, contact your Avantor sales representative.