Sterile Guinea Pig Plasma, Rockland Immunochemicals
Supplier: Rockland Immunochemical
Plasma is obtained from non-hemolyzed blood collected from healthy, fasted donors
Animals are maintained on an antibiotic-free diet. Sterilization is accomplished by Millipore filtration.
Ordering information: To specify sex and specific strain, contact your Avantor sales representative.