Nonsterile Swine Plasma, Rockland Immunochemicals
Supplier: Rockland Immunochemical
D417-04-0100 D417-06-0100 D417-04-0050 D417-06-0050
RLD417-04-0100EA 763.73 USD
RLD417-04-0100 RLD417-06-0100 RLD417-04-0050 RLD417-06-0050
Nonsterile Swine Plasma, Rockland Immunochemicals
Plasma
Plasma is obtained from non-hemolyzed blood collected from healthy, fasted donors
Animals are maintained on an antibiotic-free diet.
Ordering information: To specify sex and specific strain, contact your Avantor sales representative.