Nonsterile Chicken Plasma, Rockland Immunochemicals
Supplier: Rockland Immunochemical
Plasma is obtained from non-hemolyzed blood collected from healthy, fasted donors
Animals are maintained on an antibiotic-free diet.
Ordering information: To specify sex and specific strain, contact your Avantor sales representative.