Hamster Complement (Fresh Frozen), Rockland Immunochemicals
Supplier: Rockland Immunochemical
C306-0010 C306-0100 C306-0050
RLC306-0010EA 458.66 USD
RLC306-0010 RLC306-0100 RLC306-0050
Hamster Complement (Fresh Frozen), Rockland Immunochemicals
Sera (Serum)
Complement Fresh Frozen
  • prepared by removal of fibrinogen through clot formation under proper temperature conditions
  • Complement is preservative FREE
Special processing techniques are used to yield products with high complement activity and low background cytotoxicity. Hamster Complement is suitable for CFT and SRH.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Immunoelectrophoresis: normal