Mouse Complement (Fresh Frozen), Rockland Immunochemicals
Supplier: Rockland Immunochemical
C301-0050 C301-0010 C301-0100
RLC301-0050EA 2471.94 USD
RLC301-0050 RLC301-0010 RLC301-0100
Mouse Complement (Fresh Frozen), Rockland Immunochemicals
Sera (Serum)
Complement Fresh Frozen
  • prepared by removal of fibrinogen through clot formation under proper temperature conditions
  • Complement is preservative FREE
Mouse Complement is ideal for hemolytic assay where hemolysis of IgG antibodies are required.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal