Hamster Complement, Rockland Immunochemicals
Supplier: Rockland Immunochemical
C206-0005
RLC206-0005EA 258.06 USD
RLC206-0005
Hamster Complement, Rockland Immunochemicals
Sera (Serum)
Complement Lyophilized with DILUENT
  • suitable for complement fixation tests (CFT) and Single radial haemolysis (SRH)
Special processing techniques are used to yield products with high complement activity and low background cytotoxicity.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Immunoelectrophoresis: normal