Rabbit 'C' (5-6 Lbs.) Complement, Rockland Immunochemicals
Supplier: Rockland Immunochemical
C204-0005
RLC204-0005EA 166.12 USD
RLC204-0005
Rabbit 'C' (5-6 Lbs.) Complement, Rockland Immunochemicals
Sera (Serum)
Complement Lyophilized with DILUENT
  • suitable for complement fixation tests (CFT) and Single radial haemolysis (SRH)
Special processing techniques are used to yield products with high complement activity and low background cytotoxicity.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Immunoelectrophoresis: normal