Tissue Powders for Adsorption, Rockland Immunochemicals
Supplier: Rockland Immunochemical
C101
C113
C120
C122
C103
C115
C116
C123
C124
C117
C119
C102
C121
RLC100EA
373.26
USD
RLC100
RLC101
RLC113
RLC120
RLC122
RLC103
RLC105
RLC106
RLC107
RLC109
RLC111
RLC114
RLC115
RLC116
RLC118
RLC123
RLC124
RLC108
RLC112
RLC117
RLC119
RLC102
RLC104
RLC110
RLC121
Tissue Powders for Adsorption, Rockland Immunochemicals
Media for Microbiology
Dehydrated Media for Microbiology
Use for protocols that require a high nuclear protein content
Powders are produced by washing the various organs repeatedly in saline to remove blood, precipitated in acetone, and micronized to a fine powder. Keep container sealed and store at 2–8°C (36–46°F). All vials contain 1g of powder.