Bovine Tissue, Rockland Immunochemicals
Supplier: Rockland Immunochemical
BV-T259 BV-T263 BV-T266 BV-T278 BV-T279 BV-T294 BV-T260 BV-T281 BV-T261 BV-T262 BV-T267 BV-T272 BV-T273 BV-T275 BV-T288 BV-T290 BV-T292 BV-T293 BV-T270 BV-T283 BV-T291 BV-T295
RLBV-T259EA 101.34 USD
RLBV-T259 RLBV-T263 RLBV-T266 RLBV-T278 RLBV-T279 RLBV-T294 RLBV-T260 RLBV-T281 RLBV-T261 RLBV-T262 RLBV-T267 RLBV-T272 RLBV-T273 RLBV-T275 RLBV-T288 RLBV-T290 RLBV-T292 RLBV-T293 RLBV-T270 RLBV-T283 RLBV-T291 RLBV-T295
Bovine Tissue, Rockland Immunochemicals
Specimens Preserved Specimens Preserved Animal Materials
Used for DNA and RNA isolation.
  • Collected from antibiotic-free animals
  • DNA/RNA isolation
All tissues are collected from normal, healthy, mature animals that have been maintained on an antibiotic free diet. Tissues are stored at −20°C or colder prior to use.
Ordering information: For specific breed, sex, or weight requirements, please contact your Avantor representative.