Bovine Serum Albumin (BSA), Fraction V
Supplier: Rockland Immunochemical
BSA-30
RLBSA-30EA 652.99 USD
RLBSA-30
Bovine Serum Albumin (BSA), Fraction V
Proteins and Peptides
The BSA is suitable for use in protease sensitive assays such as RIA, EIA, and nucleic acid hybridization. It is also used to remove non-specific binding or to stabilize proteins and enzymes. BSA, Fraction V is a 30% solution in 0.85% sodium chloride with 1% octanoic acid.
  • Preservative- and stabilizer-free.