Whole Horse Sera, Rockland Immunochemicals
Supplier: Rockland Immunochemical
B307
RLB307EA 245 USD
RLB307
Whole Horse Sera, Rockland Immunochemicals
Sera (Serum)
The whole sera are prepared from normal sera by a multi-step process that includes delipidation and selective precipitation
All sera are derived from starting materials of North American origin and collection was performed in USDA inspected facilities. All materials have been inspected and certified to be free of disease and are suitable for exportation.