Anti-IgG Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
807-4602
RL807-4602EA 283.13 USD
RL807-4602
Anti-IgG Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Golden Syrian Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR