Anti-IgG Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
806-1102
RL806-1102EA 194.33 USD
RL806-1102
Anti-IgG Goat Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR