Anti-IgG Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
803-1602
RL803-1602EA 283.13 USD
RL803-1602
Anti-IgG Goat Polyclonal Antibody (Biotin)
Antibodies
Suitable for immunoblotting, ELISA, immunochemistry, immunomicroscopy as well as other antibody based assays using streptavidin or avidin conjugates requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer specificity.

Recommended Dilutions: ELISA: 1:80,000-1:800,000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR