Anti-IgG Goat Polyclonal Antibody (Texas Red®)

Supplier: Rockland Immunochemical
720-109-440
RL720-109-440EA 292.54 USD
RL720-109-440
Anti-IgG Goat Polyclonal Antibody (Texas Red®)
Antibodies
Recommended Dilutions:
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Armenian, Golden Syrian Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR